Anti-Talin 2 Antibody
BSBTPB9961
- Antibody type:Primary
- Antigen name:Talin 2
- Antigen symbol:TLN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:83660
- Antigen synonyms:ILWEQ
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Talin-2(TLN2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: TLN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat