In-Gel tryptic digestion kit, Pierce™
Catalog # PIER89871
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 KIT
- Description:In-Gel tryptic digestion kit, 150 reactions
- Product type:Peptides
- For use with:Mass spectrometer
Specifications
About this item
The Pierce™ In-Gel Tryptic Digestion Kit provides a complete set of reagents for digesting and recovering peptide fragments from approximately 150 stained protein bands excised from SDS-PAGE (polyacrylamide gel plugs).
- Includes all necessary reagents for destaining Coomassie®- or fluorescent dye-stained proteins, reduction and alkylation of cystines, and tryptic digestion
- Procedure and reagents produce reliable digestions and data generation using a wide range of conditions without optimisation
- Contains highly purified and modified MS-grade trypsin that shows no chymotryptic activity and minimal autolytic activity
Delivery: The kit includes modified porcine trypsin, destaining buffers, reduction reagents, alkylation reagents and digestion buffers along with detailed and simple instructions.