Classic magnetic IP/Co-IP kit, Pierce™
Catalog # PIER88804
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 KIT
- Description:Classic Magnetic IP/Co-IP Kit, sufficient for 40 IP reactions using 25 µl of beads
Specifications
About this item
The Pierce™ Classic Magnetic IP/Co-IP Kit enables highly effective and efficient immunoprecipitation (IP) and co-immunoprecipitation (co-IP) of antigens and protein-complexes using less than 10 µg of antibody and a magnetic separator.
- No leaching of protein A/G in the presence of detergents, low pH buffers or common mass spectrometry solvents
- Immunoprecipitate in as little as 30 minutes to reduce background and improve the capture of transient protein complexes
- Efficient and convenient with all essential buffers to complete immunoprecipitation experiments
- Magnetic beads based on Protein A/G recombinant protein ensures compatibility with most primary antibodies
Delivery: The kit contains Protein A/G magnetic beads, 1 ml; IP lysis/wash buffer, 2×50 ml; lane marker sample buffer (5X), 5 ml; elution buffer, 5 ml; and neutralisation buffer, 0,5 ml.