Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Phe-Arg beta-naphthylamide dihydrochloride
Phe-Arg β-naphthylamide dihydrochloride, powder
Phe-Arg β-naphthylamide dihydrochloride, powder
:
MP Biomedicals
Phe-Arg β-naphthylamide dihydrochloride, powder
:
MP Biomedicals
:
100929-99-5
:
:
C₂₅H₃₂Cl₂N₆O₂
:
MFCD00058046
:
519.471 g/mol