Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Melanin
Melanin
Melanin
Melanin
Supplier:
MP Biomedicals
Certificates
Expand image
Melanin
Supplier:
MP Biomedicals
About this item
CAS Number:
8049-97-6
:
Formula:
C₁₈H₁₀N₂O₄
MDL Number:
MFCD00131581
:
318.288 g/mol
Specification Test Results
Form:
Dark Gray Powder
Storage:
Store at −0 °C.
Source:
Synthetic
Specification Test Results