Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
α-Linolenic acid
α-Linolenic acid 45 - 65%, liquid
α-Linolenic acid 45 - 65%, liquid
α-Linolenic acid 45 - 65%, liquid
Supplier:
MP Biomedicals
Certificates
Expand image
α-Linolenic acid 45 - 65%, liquid
Supplier:
MP Biomedicals
About this item
CAS Number:
463-40-1
:
Formula:
C₁₈H₃₀O₂
MDL Number:
MFCD00065720
:
278.44 g/mol