Calcium hydroxide, powder USP, Ph. Eur., BP, JP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:1305-62-0
:
Formula:Ca(OH)₂
MDL Number:MFCD00010901
:74.09 g/mol
Merck Index:13,01675
Melting Pt:550 °C
Density:2.24 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-137-3
UN:3262
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Assay:95% Min.