
Orthoboric acid, powder NF
Catalog # SPCMBO110-12KG
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Poly pail
- Pk:12 kg
- Seq:4187
Specifications
Specification Test Results
- Assay:99.5% Min.
Specification Test Results
About this item
:
Merck Index:13,01326
Boiling Pt:185 °C (1013 hPa)
Melting Pt:160 °C
Density:1.44 g/cm³ (25 °C)
:61.83 g/mol
Storage Temperature:Ambient
EINECS:233-139-2
REACH:01-2119486683-25
:GHS08
CAS Number:10043-35-3
Formula:B(OH)₃
MDL Number:MFCD00011337