Order Entry
Norway
ContactUsLinkComponent
Cholesterol NF
Cholesterol NF
Supplier:  Spectrum Chemical
Certificates
Cholesterol NF
Supplier:  Spectrum Chemical

Specification Test Results

  • ASSAY (DRIED BASIS):
    95.0 - 102.0 %
  • MELTING RANGE:
    147° - 150°C
  • SPECIFIC ROTATION:
    -34° to -38°
  • SOLUBILITY IN ALCOHOL:
    No deposit or turbidity forms
  • ACIDITY:
    max 0.3 ml
  • LOSS ON DRYING:
    max 0.3%
  • RESIDUE ON IGNITION:
    max 0.1%
  • β-CHOLESTANOL (5a-CHOLESTAN-3β-ol,DIHYDROCHOLESTEROL):
    max 0.6 %
  • DESMOSTEROL:
    max 4%
  • LATHOSTEROL:
    max 2%
  • 24-DEHYDROLATHOSTEROL (5a-CHOLESTA-7,24 DIEN--3β-ol ):
    max 0.2 %
  • 4-METHYLCHOLEST-5-EN-3β-OL:
    max 0.5 %
  • TOTAL IMPURITIES INCLUDING RELATED STEROLS:
    max 5%
  • ELEMENTAL IMPURITIES:
    AS REPORTED
  • IDENTIFICATION (A):
    SPECTRUM MATCHES REFERENCE
  • IDENTIFICATION (B):
    -34° to -38°
  • IDENTIFICATION (C):
    147 - 150 °C

Specification Test Results