


2-Cyanoethyl-N,N-diisopropylchlorophosphoramidite 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:89992-70-1
:
Formula:C₉H₁₈ClN₂OP
MDL Number:MFCD00011544
:236.68 g/mol
Boiling Pt:103…105 °C (0.08 mmHg)
Density:1.061
Flash Pt:>110 °C (230 °F)
ADR:4.2,I
:GHS02
:GHS05
:GHS07