Order Entry
Norway
ContactUsLinkComponent
Linear polyacrylamide solution 5 mg/ml for molecular biology, for DNA/RNA precipitation
Linear polyacrylamide solution 5 mg/ml for molecular biology, for DNA/RNA precipitation
Catalog # J67830.XF
CAS Number:  
Linear polyacrylamide solution 5 mg/ml for molecular biology, for DNA/RNA precipitation
Catalog # J67830.XF
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
Storage Temperature:Freezer