Hyaluronic acid sodium salt (from Streptococcus equi) 91%
Catalog # J66993.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Merck Index:14,4757
Storage Temperature:Freezer
CAS Number:9067-32-7
Formula:(C₁₄H₂₁NaNO₁₁)n
MDL Number:MFCD00875848