Order Entry
Norway
ContactUsLinkComponent
Glycine, 0.2M buffer soln., pH 2.5
Glycine, 0.2M buffer soln., pH 2.5
Certificates
Glycine, 0.2M buffer soln., pH 2.5
Certificates

Specifications

  • CAS No.:
    56-40-6
  • MDL No.:
    MFCD00008131
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless

Specifications

Related Information