Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Adenosine-3',5'-cyclic monophosphate (cAMP)
Adenosine-3',5'-cyclic monophosphate (cAMP) 98%
Adenosine-3',5'-cyclic monophosphate (cAMP) 98%
:
Thermo Fisher Scientific
Adenosine-3',5'-cyclic monophosphate (cAMP) 98%
:
Thermo Fisher Scientific
:
60-92-4
:
:
C₁₀H₁₂N₅O₆P
:
MFCD00005845
:
329.21 g/mol
:
14,2708
:
260 °C (dec.)
:
200-492-9
msds_ALFAJ60936
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years