Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fmoc-Glu(OAll)-OH
Fmoc-Glu(OAll)-OH 95%
Fmoc-Glu(OAll)-OH 95%
Fmoc-Glu(OAll)-OH 95%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Fmoc-Glu(OAll)-OH 95%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
133464-46-7
:
Formula:
C₂₃H₂₃NO₆
MDL Number:
MFCD00190879
:
409.438 g/mol
Related Information
Documentation
Certificates
Search by Lot Number
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation