4-Methoxy-2-methylbenzaldehyde 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:52289-54-0
:
Formula:C₉H₁₀O₂
MDL Number:MFCD02261771
:150.18 g/mol
Boiling Pt:262…267 °C (lit.)
Density:1.107
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient