Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fmoc-Inp-OH
Fmoc-Inp-OH 98%
Fmoc-Inp-OH 98%
Fmoc-Inp-OH 98%
Leverandør:
Thermo Fisher Scientific
Sertifikat
Expand image
Fmoc-Inp-OH 98%
Leverandør:
Thermo Fisher Scientific
Om dette produktet
CAS-nummer:
148928-15-8
:
Formel:
C₂₁H₂₁NO₄
MDL-nummer:
MFCD00273475
:
351.402 g/mol
Molten Pt:
∼ 190 °C (dec.)
Relatert informasjon
Documentation
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation