Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-5-phenyl-1,3,4-thiadiazole
2-Amino-5-phenyl-1,3,4-thiadiazole 97%
2-Amino-5-phenyl-1,3,4-thiadiazole 97%
H32826.03
:
Thermo Fisher Scientific
:
2-Amino-5-phenyl-1,3,4-thiadiazole 97%
H32826.03
:
Thermo Fisher Scientific
:
Pk:
1 g
Seq:
3047
:
2002-03-1
:
:
C₈H₇N₃S
:
MFCD00205278
:
177.23 g/mol
:
227…230 °C
:
GHS07
:
Ambient