Order Entry
Norway
ContactUsLinkComponent
Dimethyl-4-aminothiophene-2,3-dicarboxylate hydrochloride 97%
Irritant
Dimethyl-4-aminothiophene-2,3-dicarboxylate hydrochloride 97%
Catalog # B20028.03
CAS Number:  
Irritant
Dimethyl-4-aminothiophene-2,3-dicarboxylate hydrochloride 97%
Catalog # B20028.03
CAS Number:  

Specifications

  • Pk:
    1 g

Specifications

About this item

CAS Number:121071-71-4
 :
Formula:C₈H₁₀ClNO₄S
 :251.681 g/mol
Melting Pt:166…169 °C
 :GHS07
Storage Temperature:Ambient
MDL Number:MFCD00205189