Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Eosin Y (yellowish)
Eosin Y (yellowish)
Eosin Y (yellowish)
Eosin Y (yellowish)
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Eosin Y (yellowish)
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
17372-87-1
:
Formula:
C₂₀H₆Br₄Na₂O₅
MDL Number:
MFCD00005040
:
691.86 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAA18578
Videos
Thermo Scientific Acroseal 25 Years
Documentation