
Methyl-3,5-di-tert-butyl-4-hydroxybenzoate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2511-22-0
:
Formula:C₁₆H₂₄O₃
MDL Number:MFCD00017253
:264.37 g/mol
Melting Pt:164…166 °C
EINECS:219-729-2
:GHS07
Storage Temperature:Ambient