kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Cinnamamide
Cinnamamide predominantly trans 97%
Cinnamamide predominantly trans 97%
:
Thermo Fisher Scientific
Cinnamamide predominantly trans 97%
:
Thermo Fisher Scientific
:
621-79-4
:
:
C₉H₉NO
:
MFCD00008033
:
147.177 g/mol
:
145…149 °C
:
210-707-8
:
GHS07
:
Ambient
msds_ALFAA13429
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years