Aluminium oxide, Drysphere®, without indicator
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1344-28-1
:11092-32-3
Formula:Al₂O₃
MDL Number:MFCD00003424
:101.96 g/mol
Merck Index:14,356
Boiling Pt:2980 °C
Melting Pt:2045 °C
Storage Temperature:Ambient
EINECS:215-691-6