Order Entry
Norway
ContactUsLinkComponent
D-Glucose dehydrogenase
D-Glucose dehydrogenase
  45402.4I
 :  
D-Glucose dehydrogenase
  45402.4I
 :  

 

  • Pk:
    1 KU
  • CAS No.:
    9028-53-9
  • Form:
    Pale yellow powder
  • MDL No.:
    MFCD00131181
  • Enzyme name:
    D-Glucose dehydrogenase

 

 

Applications: Diagnostics and cofactor regeneration of NAD(P)H in reductive processes.

Physical Form: Powder Appearance: Pale yellow MDL No.: MFCD00131181

Applications: Diagnostics and cofactor regeneration of NAD(P)H in reductive processes