Starch, soluble modified (insolubles 0.01% max)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9005-84-9
:
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00082026
:162.14 g/mol
Merck Index:13,08878
Storage Temperature:Ambient
EINECS:232-686-4