Order Entry
Norway
ContactUsLinkComponent
Poly(styrene), test film
Poly(styrene), test film
Catalog # 44502.KT
CAS Number:  
Poly(styrene), test film
Catalog # 44502.KT
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :125000 - 250000 g/mol
Storage Temperature:Ambient
CAS Number:9003-53-6
Formula:C₈H₈
MDL Number:MFCD00084450