Order Entry
Norway
ContactUsLinkComponent
 
Ground & Waste Water Pollution Standard Solution 1, Specpure®
Catalog # 36740.AE
CAS Number:  
Ground & Waste Water Pollution Standard Solution 1, Specpure®
Catalog # 36740.AE
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pk:
    100 mL
  • Description:
    Ground & Waste Water Pollution Standard Solution 1
  • MDL No.:
    MFCD02091711
  • UN No.:
    3264

Specifications

About this item

For US EPA Clean Water Act.

  • 15 Analytes

UN No.: UN3264
MDL No.: MFCD02091711