Order Entry
Norway
ContactUsLinkComponent
 
Grinding pieces, ceramic, Al-23
  33190.A1
Grinding pieces, ceramic, Al-23
  33190.A1

 

  • Ø:
    10 mm
  • Description:
    Grinding pieces, Al-23, 10 mm, 440 pieces/kg
  • Length:
    10 mm
  • Material:
    Ceramic
  • Seq:
    01
  • Pk:
    1 kg