Anti-CD46 Rabbit Polyclonal Antibody
Catalog # BSBTPB9848
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:CD46 antigen, complement regulatory protein
- Antigen symbol:CD46
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:17221
- Antigen synonyms:Mcp
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CD46
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse