Apron, Vinyl
113-0237
: Ansell
- Klærstørrelse:L
- Antrekksapplikasjoner:Chemical protection
- Farge:Blå
- Seq:03
- Fp:1
- Tykkelse:0,2 mm
This disposable, chemical resistant apron is made of virgin vinyl. It provides excellent protection when handling acids and caustic solutions.
- Heat-sealed binding at the neck and bottom eliminates leaks caused by exposed elastic and thread
- Back closure consists of two plastic snaps and nylon ties
- No exposed elastic or metal parts to create static and corrosion
- Full-length sleeves provide additional coverage
Ideal for use in chemical processing, oil refining, petrochemicals, beef/pork/poultry/fish processing, janitorial and maintenance work and for use in cleanrooms.
Colour: Blue
: Food contact approved.