Anti-TBL1XR1 Rabbit Polyclonal Antibody
ABGEAP16411A
: Abgent
- Antibody type:Primary
- Antigen name:transducin (beta)-like 1 X-linked receptor 1
- Antigen symbol:TBL1XR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Gene ID:79718
- Antigen synonyms:C21|DC42|TBLR1|IRA1
- Amino acid number:109 to 137
- Storage buffer:pH 7,4
- Molecular weight:55.595 kDa
- Sequence:AKNGENTANGEENGC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.4
- Shipping temperature:4 °C
- Immunogen:This TBL1XR1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 109-137 amino acids from the N-terminal region of human TBL1XR1.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: TBL1XR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human