ESD safety shoes, lace-up, SK Grip, Electro Grip
: U-Power
ESD safety shoes made from water repellent full grained leather and a lightweight, extremely flexible, slip resistant PU/PU U-Grip sole.
- Anatomically shaped Polysoft insole made from soft PU, with pockets in the foot support area
- PU/PU walking sole with 90° angle blocks and multilevel stabilising channels to for extremely high anti-slip features; TPU anti twist insert for additional support
- Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
- Stress-out tongue system allows easy instep and provides added comfort when walking
- Natural fit width 11 for additional comfort
- Air toe protective aluminium toecap with breathable membrane (200 J)
- Light, flexible and non metallic, puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Colour: Black
: EN 61340 (ESD); EN 20345:2011; S3, SRC