Order Entry
Norway
ContactUsLinkComponent
 
Cerium(IV) sulphate 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Katalog # 5626-02
Supplier:  Avantor
CAS Number:  
undefined
Cerium(IV) sulphate 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Katalog # 5626-02
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE bottle, narrow mouth
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    0.095 - 0.105
  • Normality:
    0.095 - 0.105

Specification Test Results

About this item

 :
 :332.24 g/mol
Merck Index:12,02036
Melting Pt:195 °C
Storage Temperature:Ambient
EINECS:237-029-5
UN:3264
ADR:8,II
 :GHS07
 :GHS05
 :GHS03
 :GHS08
CAS Number:13590-82-4
Formula:Ce(SO₄)₂
MDL Number:MFCD00148852