Order Entry
Norway
ContactUsLinkComponent
 
 
 
Potassium hydroxide 85.0-100.5%, pellets NF, FCC, Macron Fine Chemicals™
 
undefined
 
 
Potassium hydroxide 85.0-100.5%, pellets NF, FCC, Macron Fine Chemicals™
 

 

  • Meets F.C.C. Requirements:
  • Meets N.F. Requirements:
  • GMP Manufactured Product:
  • Food GMP Manufactured Product:
  • Storage: Preserve in tight containers.:
    = 0.002 %
  • CAUTION: For Manufacturing, processing or repackaging:
    85.0 - 100.5 %
  • Bulk Pharmaceutical Chemical:
    = 85.0 %
  • Bulk Food Chemical:
    = 3.5 %
  • NF - Assay (Total Alkali):
    ≥85.0 %
  • NF - Carbonate (as K₂CO₃):
    ≤3.5 %
  • NF - Content of Potassium:
    ≥59.9 %
  • NF - Limit of Sodium:
    ≤1.0 %
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Insoluble Substances:
    Passes Test
  • FCC - Assay (KOH):
    85.0 - 100.5 %
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Insoluble Substances:
    Passes Test
  • FCC - Lead (Pb):
    ≤2 mg/kg
  • FCC - Mercury (Hg):
    ≤0.1 ppm
  • FCC - Potassium Carbonate (K₂CO₃):
    ≤3.5 %
  • Solubility:
    Passes Test

 

 

Frequently Bought Together