VWR® La-Pha-Pack™ ProFill, Syringe filters
Supplier: LA PHA PACK
Certificates
About this item
The ProFill HPLC syringe filters are suitable for sample volumes of 1,5 ml up to 100 ml. The filters have a PTFE membrane and a PP housing.
The membrane type and pore size are printed onto the PE-bag, in which the filters are tamper-proof evidently packed.
Sample retention: <0,1 ml
Packaging: The membranes are packed per 100 in a tamper-evident PE bag. Membrane and pore size are printed onto each bag. An additional blue reclosable PP box offers further protection for the filters during transportation.