Order Entry
Norway
ContactUsLinkComponent
Safety Storage Cabinet, S-CLASSIC-90
Safety Storage Cabinet, S-CLASSIC-90
Catalog # 135-2678
Supplier:  asecos
undefined
Safety Storage Cabinet, S-CLASSIC-90
Catalog # 135-2678
Supplier:  asecos

Specifications

  • Description:
    Tall cabinet type 90, S-CLASSIC-90, model S90.196.120.MV.WDAS without interior equipment (sold separately), body light grey / doors light grey (RAL 7035)
  • Seq:
    01
  • Doors:
    2
  • Pk:
    1
  • Weight:
    515 kg
  • W×D×H ext.:
    1196×616×1968 mm

Specifications

About this item

Fireproof safety cabinets, classified type 90 (fire resistant up to 90 minutes), safe and approved storage of flammable hazardous substances in working areas.

  • Robust construction and longevity - triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
  • Two in one - 2 compartments (vertical separation) allow the storage of flammable liquids and aggressive chemicals in one cabinet
  • User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
  • No unauthorised use - doors lockable with profile cylinder (integration in an existing locking system possible)
  • Easy alignment - adjusting aids to compensate for uneven floor
  • Ventilation - integrated air ducts ready for connection (DN 75) to a technical exhaust system
Certifications: Furnace tested (type test) in accordance with EN 14470-1
Ordering information: Interior equipment packages need to be ordered separately (not included). Please check the website or contact Avantor for further information.
Delivery: Delivery prices available upon request.