Order Entry
Norway
ContactUsLinkComponent
 
 
2-Acetamidofluorene for synthesis
  8.20576.0010
 :  Merck
 :  
 
2-Acetamidofluorene for synthesis
  8.20576.0010
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

 

 

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    190 - 193 °C
  • Molar Mass:
    223.27 g/mol
  • Storage temperature:
    +15...+25 °C
  • Identity (IR):
    Passes test

 

 

 :
 :223.28 g/mol
 :M-Clarity™ quality level = MQ200
 :200-188-6
 :GHS08
 :Ambient
 :53-96-3
 :C₁₅H₁₃NO
 :MFCD00001116