Anti-MTFR1 Rabbit Polyclonal Antibody
BOSSBS-7632R
: Bioss
- Antibody type:Primary
- Antigen name:Mitochondrial Fission Regulator 1
- Antigen symbol:MTFR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:9650
- Antigen synonyms:Chondrocyte protein with a poly proline region|Mitochondrial fission regulator 1|MTFR 1|Chondrocyte protein with a poly-proline region|KIAA0009|MTFR1_HUMAN.|CHPPR|FAM54A2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
This gene encodes a mitochondrial protein that is characterized by a poly-proline rich region. A chicken homolog of this protein promotes mitochondrial fission and the mouse homolog protects cells from oxidative stress. A related pseudogene of this gene is found on chromosome X.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: MTFR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat