Protectivel hood, full, Flowhood 21 (FH21)
Katalog # SCTS2023098
Supplier: SCOTT SAFETY
Specifications
- Description:Protective hood, FH21, anti-static, without hose
- Pk:1
Specifications
About this item
This headtop provides the highest levels of comfort and wearer acceptance. It is designed to be used with the following respiratory systems: Spirit, Tornado, Proflow2 120SC/120EX, 160SC/160EX and Proflow3 120EX, 180SC.
- Revolutionary soft head cradle for intuitive and straightforward donning
- Ultra-clear PETG visor giving wide field of view and a clear down-view panel
- Anti-static full hood design with skin-friendly neoprene chin seal
Certifications: EN 14941 (class TH3), EN 166 F
Ordering information: FH21 is supplied without hose.