Anti-IgG Goat Polyclonal Antibody
Catalog # IMMRIR2926
Supplier: ImmunoReagents
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Pig
- Western blot:Yes
- Cross adsorption:No
- Format:Purified IgG
- Form:Clear, colorless liquid, 0.2 µm filtered
- Storage buffer:10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2
- Storage temperature:2-8 °C
- Concentration:> 4,5 mg/ml (E 1% at 280 nm = 13,0)
- Shipping temperature:2-8 °C
- Immunogen:Pig IgG
- Purification:Affinity purified using solid phase Pig IgG
- Size:1,0 mg
- Pk:1 mg
Specifications
About this item
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Pig