Order Entry
Norway
Orders LinkContactUsLinkComponent
Diffraction gratings
Marketsource
Diffraction gratings
Marketsource
Diffraction gratings
Diffraction gratings
Photographic diffraction objects in slide frames.
  • Dimensions: 50×50 mm
Product Family Options
  • Description
  • Diaphragm with 3 double slits, Slit spacing: 0,30 mm, Slit width: 0,10 mm; 0,15 mm; 0,20 mm
    1 * 1 Items
    1 * 1 Items - kr 335,00
    kr 335,00
    1 * 1 Items
       kr 335,00
    Marketsource
    Marketsource
  • Diaphragm with 4 double slits, Slit spacing: 0,15 mm, Slit width: 0,25 mm; 0,50 mm; 0,75 mm; 1,00 mm
    1 * 1 Items
    1 * 1 Items - kr 335,00
    kr 335,00
    1 * 1 Items
       kr 335,00
    Marketsource
    Marketsource
  • Diaphragm with 4 multiple slits, Slit spacing: 0,25 mm, Slit width: 0, 15 mm
    1 * 1 Items
    1 * 1 Items - kr 335,00
    kr 335,00
    1 * 1 Items
       kr 335,00
    Marketsource
    Marketsource