Order Entry
Norway
Orders LinkContactUsLinkComponent
Tuning Fork

Tuning Fork

Tuning fork, length 105 mm, 1700 Hz
Catalog # BINHU10115
Marketsource
Supplier:   3B Scientific
Tuning Fork
Catalog # BINHU10115
Supplier:  3B Scientific
Marketsource
Tuning Fork
Tuning Fork

Tuning fork, length 105 mm, 1700 Hz

Price
kr 1 100,00
Log in for availability
1 * 1 Items
Product Details & Documents

Suitable as a source of intense, high frequency sound, for example, for producing stationary sound waves in Kundt's tube.

Specifications
  • Description:
    Tuning fork, length 105 mm
  • Frequency:
    1700 Hz
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.