Order Entry
Norway
ContactUsLinkComponent
Anti-CD2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: RPA-2.10]
Anti-CD2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: RPA-2.10]
Supplier:  Tonbo Biosciences
Certificates
Anti-CD2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: RPA-2.10]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 50-0029-T025
  • 50-0029-T100
  • 50-0029-T500
  • Antigen symbol:
  • CD2
  • CD2
  • CD2
  • Antigen name:
  • CD2 molecule
  • CD2 molecule
  • CD2 molecule
  • Antigen synonyms:
  • sheep red blood cell receptor|T-cell surface antigen T11|erythrocyte receptor|rosette receptor|T11|Leu-5|SRBC|LFA-3 receptor|lymphocyte-function antigen-2|LFA-2
  • sheep red blood cell receptor|T-cell surface antigen T11|erythrocyte receptor|rosette receptor|T11|Leu-5|SRBC|LFA-3 receptor|lymphocyte-function antigen-2|LFA-2
  • sheep red blood cell receptor|T-cell surface antigen T11|erythrocyte receptor|rosette receptor|T11|Leu-5|SRBC|LFA-3 receptor|lymphocyte-function antigen-2|LFA-2
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • RPA-2.10
  • RPA-2.10
  • RPA-2.10
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 914
  • 914
  • 914
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Cross adsorption:
  • No
  • No
  • No
  • Storage temperature:
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.

Specifications