Cryogenic vials, glass
: Hycultec
Glass vials, with screw cap. These vials are suitable for −20 °C deepfreezing. Screw caps with silicone stoppers make sure that the reagents are stored tight and secure.
- Available with aluminium / coloured plastic / perforated plastic screw caps