Anti-DCXR Antibody Pair
ABNOH00051181-AP51
: Abnova
- Antigen name:Dicarbonyl/L-xylulose reductase
- Antigen symbol:DCXR
- Host:Multiple
- Reactivity:Human
- Gene ID:51181
- Antigen synonyms:PNTSU|Kidney dicarbonyl reductase|Carbonyl reductase II|kiDCR|HCR2|SDR20C1|KIDCR|Sperm surface protein P34H|XR|L-xylulose reductase|HCRII|P34H|Dicarbonyl/L-xylulose reductase|DCR
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
This Set comes with matched Antibody Pair to detect and quantify protein level of human DCXR.
Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-DCXR (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-DCXR (50 µg);
Type:
Antigen: DCXR
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human