Anti-PRKAG2 Polyclonal Antibody Pair
Catalog # ABNOH00051422-PW1
Supplier: Abnova
Specifications
- Antigen name:Protein Kinase, AMP-activated, Non-catalytic Subunit Gamma 2
- Antigen symbol:PRKAG2
- Clonality:Polyclonal
- Host:Multiple
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:51422
- Antigen synonyms:CMH6|AAKG|AAKG2|WPWS|H91620p
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-PRKAG2 (300 µl); 2. Antibody pair for Western Blot: Mouse Polyclonal Anti-PRKAG2 (50 µl)
Type:
Antigen: PRKAG2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human