Anti-PARK2 + GRIN2B Antibody Pair
Catalog # ABNODI0233
Supplier: Abnova
Specifications
- Antigen name:Parkin And Glutamate Receptor, Ionotropic, N-Methyl D-Aspartate 2B
- Antigen symbol:PARK2 + GRIN2B
- Host:Multiple
- ImmunoFluorescence:Yes
- Gene ID:5071|2904
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the PARK2 protein, and the other against the GRIN2B protein for use in Proximity Ligation Assay
Antibody pair set content: 1. PARK2 Rabbit purified Polyclonal Antibody (20 µg) 2. GRIN2B Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Type:
Antigen: PARK2 + GRIN2B
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: