Anti-ASPA Antibody Pair
ABNOH00000443-AP11
: Abnova
- Antigen name:Aspartoacylase
- Antigen symbol:ASPA
- Host:Multiple
- Reactivity:Human
- Gene ID:443
- Antigen synonyms:ASPA, ACY2, ASP, Aspartoacylase, Aminoacylase-2
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
This Set comes with matched Antibody Pair to detect and quantify protein level of human ASPA.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-ASPA (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-ASPA, IgG2b kappa (20 µg);
Type:
Antigen: ASPA
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human