Order Entry
Norway
ContactUsLinkComponent
Anti-IL10 + A2M Antibody Pair
Anti-IL10 + A2M Antibody Pair
Catalog # ABNODI0037
Supplier:  Abnova
Anti-IL10 + A2M Antibody Pair
Catalog # ABNODI0037
Supplier:  Abnova

Specifications

  • Antigen symbol:
    IL10 + A2M
  • Host:
    Multiple
  • ImmunoFluorescence:
    Yes
  • Gene ID:
    3586|2
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

Specifications

About this item

This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the IL10 protein, and the other against the A2M protein for use in Proximity Ligation Assay

Antibody pair set content: 1. IL10 Rabbit purified Polyclonal Antibody (20 µg) 2. A2M Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: IL10 + A2M
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: