Order Entry
Norway
ContactUsLinkComponent
 
Anti-LCK Antibody Pair
  ABNOH00003932-AP11
 :  Abnova
Anti-LCK Antibody Pair
  ABNOH00003932-AP11
 :  Abnova

 

  • Antigen name:
    Lymphocyte-specific Protein tyrosine kinase
  • Antigen symbol:
    LCK
  • Host:
    Multiple
  • Reactivity:
    Human
  • Gene ID:
    3932
  • Antigen synonyms:
    p56lck|LSK|tyrosine-protein kinase Lck|YT16|leukocyte C-terminal Src kinase|T-lymphocyte specific protein tyrosine kinase p56lck|p56(LSTRA) protein-tyrosine kinase|RP4-675E8.4|t cell-specific protein-tyrosine kinase|lymphocyte cell-specific protein-tyrosine kinase|proto-oncogene tyrosine-protein kinase LCK|pp58lck
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

 

 

This Set comes with matched Antibody Pair to detect and quantify protein level of human LCK.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-LCK (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-LCK, IgG1 kappa (20 µg);

Type:
Antigen: LCK
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human